Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corticotropin Releasing Factor Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Corticotropin Releasing Factor |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Corticotropin Releasing Factor Polyclonal specifically detects Corticotropin Releasing Factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Corticotropin Releasing Factor | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
1392 | |
Antibody was raised against a 16 amino acid peptide near the amino terminus of human CRH. The immunogen is located within amino acids 30 - 80 of CRH. Amino Acid Squence: FQPPPQSEQPQQPQAR | |
Primary | |
CRH antibody is human specific. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human | |
corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor | |
CRH | |
IgG | |
Affinity Purified | |
29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title