Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cortisol Antibody (BGN/C53) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100624840.025MG
Description
Cortisol Monoclonal specifically detects Cortisol in All Species samples. It is validated for ELISA.Specifications
| Cortisol | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:500-1:5000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| This product recognises the glucocorticoid hormone cortisol, produced in the adrenal cortex and involved in responses to stress. Cross reactivity: Cortisol 100%, Corticosterone 50%, Prednisolone 15%, 11-Desoxycortisol, <0.1% Cortisone,<0.1% 17-a-OH Progesterone, <0.1%. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| BGN/C53 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Synthetic corticosterone conjugated to Bovine Serum Albumin. | |
| 0.025 mg | |
| Primary | |
| All species | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction