Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisol Antibody (BGN/C53) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen Cortisol
Clone BGN/C53
Concentration 1.0 mg/mL
Dilution ELISA 1:500-1:5000
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017485
SDP
View Documents
Novus Biologicals
NB100624840.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10062484
SDP
View Documents
Novus Biologicals
NB10062484
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Cortisol Monoclonal specifically detects Cortisol in All Species samples. It is validated for ELISA.
Specifications

Specifications

Cortisol
1.0 mg/mL
ELISA
Unconjugated
RUO
Synthetic corticosterone conjugated to Bovine Serum Albumin.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
BGN/C53
ELISA 1:500-1:5000
Monoclonal
Mouse
All species
IgG3
Affinity Purified
This product recognises the glucocorticoid hormone cortisol, produced in the adrenal cortex and involved in responses to stress. Cross reactivity: Cortisol 100%, Corticosterone 50%, Prednisolone 15%, 11-Desoxycortisol, <0.1% Cortisone,<0.1% 17-a-OH Progesterone, <0.1%.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.