Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cortisol Parameter Assay Kit

Catalog No. kge008b
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Cortisol Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Cortisol Parameter Assay Kit quantifies multi-species Cortisol in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (20 ul), urine (10 ul).

Specifications

Quantity 1 Kit
Species Multi-species
Conjugate Biotin
Binding Target Cortisol
Assay Sensitivity 0.111 ng/mL
Assay Range 0.16 to 10 ng/mL
For Use With (Application) ELISA
Sample Type Cell Culture Supernates (10uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Saliva (20uL), Urine (10uL)
Test Time 2.5 hr.
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.