Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COUP-TF I/NR2F1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15282520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15282520 20 μL
NBP152825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15282520 Supplier Novus Biologicals Supplier No. NBP15282520UL

Rabbit Polyclonal Antibody

COUP-TF I/NR2F1 Polyclonal specifically detects COUP-TF I/NR2F1 in Human samples. It is validated for Western Blot.

Specifications

Antigen COUP-TF I/NR2F1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P10589
Gene Alias COUP transcription factor 1, COUP-TF1, COUP-TFI, EAR-3COUP transcription factor I, EAR3COUP-TF I, ERBAL3V-erbA-related protein 3, Nuclear receptor subfamily 2 group F member 1, nuclear receptor subfamily 2, group F, member 1, SVP44, TCFCOUP1, TFCOUP1NR2F2, transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-ahomolog-like 3)
Gene Symbols NR2F1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NR2F1(nuclear receptor subfamily 2, group F, member 1) The peptide sequence was selected from the N terminal of NR2F1. Peptide sequence GEQQQQAGSGAPHTPQTPGQPGAPATPGTAGDKGQGPPGSGQSQQHIECV.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7025
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.