Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COUP-TF II/NR2F2 Rabbit anti-Human, Alexa Fluor 350, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB106326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106326 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106326 Supplier Novus Biologicals Supplier No. NBP298861AF350

Rabbit Polyclonal Antibody

COUP-TF II/NR2F2 Polyclonal specifically detects COUP-TF II/NR2F2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COUP-TF II/NR2F2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias Apolipoprotein A-I regulatory protein 1, ARP-1, ARP1ADP-ribosylation factor related protein 1, chicken ovalbumin upstream promoter transcription factor 2, COUP transcription factor 2, COUP transcription factor II, COUP-TF II, COUP-TF2, COUPTFB, COUPTFII, COUP-TFII, MGC117452, Nuclear receptor subfamily 2 group F member 2, nuclear receptor subfamily 2, group F, member 2, SVP40, TFCOUP2apolipoprotein AI regulatory protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human COUP-TF II/NR2F2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 7026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.