Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX-1 Antibody (345305), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB371556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371556 Supplier Novus Biologicals Supplier No. FAB3740K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

COX-1 Monoclonal specifically detects COX-1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen COX-1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 345305
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS
Host Species Rat
Immunogen E. coli-derived recombinant human COX-1, Ala24-Leu599, Accession # P23219
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5742
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.