Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX-1 Antibody (6B6), Novus Biologicals™
SDP

Catalog No. NB298252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298252 50 μL
NB298253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298252 Supplier Novus Biologicals Supplier No. NBP32636250UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

COX-1 Monoclonal antibody specifically detects COX-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen COX-1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 6B6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS
Host Species Rabbit
Immunogen A synthesized peptide derived from Human COX-1 [UniProt P23219]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5742
Target Species Human, Mouse
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.