Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX-1 Antibody (6B6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | COX-1 |
---|---|
Clone | 6B6 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
COX-1 Monoclonal antibody specifically detects COX-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
COX-1 | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS | |
A synthesized peptide derived from Human COX-1 [UniProt P23219] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6B6 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Hypoxia, Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5742 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title