Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX-2 Antibody (COX2/1941), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB382163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382163 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382163 Supplier Novus Biologicals Supplier No. NBP308272C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX-2 Monoclonal antibody specifically detects COX-2 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen COX-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone COX2/1941
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase)
Host Species Mouse
Immunogen Recombinant human COX-2 protein fragment (around aa 442-572) (exact sequence is proprietary) (Uniprot: P35354)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Hypoxia, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5743
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.