Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX10 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB050313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050313 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050313 Supplier Novus Biologicals Supplier No. NBP297977AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COX10 Polyclonal specifically detects COX10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen COX10
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias COX10 (yeast) homolog, cytochrome c oxidase assembly protein (heme A:farnesyltransferase), COX10 homolog, cytochrome c oxidase assembly protein, heme A:farnesyltransferase (yeast), cytochrome c oxidase assembly protein, cytochrome c oxidase subunit X, EC 2.5.1.-, heme A: farnesyltransferase, Heme O synthase, protoheme IX farnesyltransferase, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human COX10 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1352
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.