Learn More
Description
Specifications
Specifications
| Antigen | COX10 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | COX10 (yeast) homolog, cytochrome c oxidase assembly protein (heme A:farnesyltransferase), COX10 homolog, cytochrome c oxidase assembly protein, heme A:farnesyltransferase (yeast), cytochrome c oxidase assembly protein, cytochrome c oxidase subunit X, EC 2.5.1.-, heme A: farnesyltransferase, Heme O synthase, protoheme IX farnesyltransferase, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human COX10 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
