Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB339362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339362 Supplier Novus Biologicals Supplier No. NB11039115MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COX4 Polyclonal antibody specifically detects COX4 in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Insect, Opossum, Primate samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP), Immunocytochemistry, KnockDown

Specifications

Antigen COX4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human COX IV isoform 1 (within residues 1-100). [Swiss-Prot# P13073]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Loading Controls, Neuroendocrinology
Primary or Secondary Primary
Gene ID (Entrez) 1327
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.