Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4NB Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB087405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087405 Supplier Novus Biologicals Supplier No. NBP300354AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COX4NB Polyclonal specifically detects COX4NB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen COX4NB
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias C16orf2, C16orf4, chromosome 16 open reading frame 2, COX4 neighbor, COX4AL, FAM158Bchromosome 16 open reading frame 4, neighbor of COX4, NOC4family with sequence similarity 158, member B, Protein FAM158B
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant COX4NB (O43402; Met1-Cys210).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.