Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX5A Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB131956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB131956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB131956 Supplier Novus Biologicals Supplier No. NBP306072AF647

Rabbit Polyclonal Antibody

COX5A Polyclonal specifically detects COX5A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen COX5A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias COX, COX-VA, Cytochrome c oxidase polypeptide Va, cytochrome c oxidase polypeptide, mitochondrial, cytochrome c oxidase subunit 5A, mitochondrial, cytochrome c oxidase subunit Va, mitochondrial cytochrome c oxidase subunit Va, VA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human COX5A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9377
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.