Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX5b Antibody (021), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB038769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038769 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038769 Supplier Novus Biologicals Supplier No. NBP290312AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

COX5b Monoclonal specifically detects COX5b in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen COX5b
Applications Western Blot, ELISA
Classification Monoclonal
Clone 21
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias COXVB, Cytochrome c oxidase polypeptide Vb, cytochrome c oxidase polypeptide VB, mitochondrial, cytochrome c oxidase subunit 5B, mitochondrial, cytochrome c oxidase subunit Vb
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human COX5b (P10606; Ala32-His129).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1329
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.