Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX5b Rabbit anti-Human, Clone: 31, Novus Biologicals™
SDP

Catalog No. NB022091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022091 100 μg
NB022092 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022091 Supplier Novus Biologicals Supplier No. NBP290314100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

COX5b Monoclonal specifically detects COX5b in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen COX5b
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 31
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias COXVB, Cytochrome c oxidase polypeptide Vb, cytochrome c oxidase polypeptide VB, mitochondrial, cytochrome c oxidase subunit 5B, mitochondrial, cytochrome c oxidase subunit Vb
Host Species Rabbit
Immunogen Recombinant Human COX5b Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1329
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.