Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6A1 Antibody (OTI4G2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP270427G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270427G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270427G Supplier Novus Biologicals Supplier No. NBP270427G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6A1 Monoclonal specifically detects COX6A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COX6A1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4G2
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias COX VIa-L, COX6A, COX6AL, Cytochrome c oxidase polypeptide VIa-liver, cytochrome c oxidase subunit 6A1, mitochondrial, cytochrome C oxidase subunit VIa homolog, cytochrome c oxidase subunit VIa polypeptide 1, Cytochrome c oxidase subunit VIA-liver, MGC104500
Gene Symbols COX6A1
Host Species Mouse
Immunogen Full length recombinant protein of human COX6A1 (NP_004364) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1337.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.