Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6A1 Monoclonal Antibody (OTI4G2), TrueMAB™, OriGene
SDP

Catalog No. p-200010400
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2289 30 μL
50-167-2575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2289 Supplier OriGene Supplier No. TA501431S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6A1 Monoclonal antibody specifically detects COX6A1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen COX6A1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4G2
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pex5
Gene Accession No. P50542, Q2M2R8
Gene Alias CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782
Host Species Mouse
Immunogen Full length human recombinant protein of human PEX5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312703, 486710, 5830
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.