Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6B2 Antibody (OTI1F5), Biotin, Novus Biologicals™
SDP

Catalog No. NBP272379B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272379B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272379B Supplier Novus Biologicals Supplier No. NBP272379B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6B2 Monoclonal specifically detects COX6B2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COX6B2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F5
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cancer/testis antigen 59, COX VIb-2, COXVIB2, CT59MGC119094, cytochrome c oxidase subunit 6B2, Cytochrome c oxidase subunit VIb isoform 2, cytochrome c oxidase subunit VIb polypeptide 2 (testis), cytochrome c oxidase subunit VIb, testes specific, cytochrome c oxidase subunit VIb, testes-specific, Cytochrome c oxidase subunit VIb, testis-specific isoform, FLJ32865, FLJ46422
Gene Symbols COX6B2
Host Species Mouse
Immunogen Full length recombinant protein of human COX6B2 (NP_653214) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 125965.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.