Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX6B2 Antibody (OTI1F5), Novus Biologicals™
SDP

Catalog No. NBP202729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202729 Supplier Novus Biologicals Supplier No. NBP202729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COX6B2 Monoclonal specifically detects COX6B2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COX6B2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F5
Concentration 0.84 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100
Gene Alias Cancer/testis antigen 59, COX VIb-2, COXVIB2, CT59MGC119094, cytochrome c oxidase subunit 6B2, Cytochrome c oxidase subunit VIb isoform 2, cytochrome c oxidase subunit VIb polypeptide 2 (testis), cytochrome c oxidase subunit VIb, testes specific, cytochrome c oxidase subunit VIb, testes-specific, Cytochrome c oxidase subunit VIb, testis-specific isoform, FLJ32865, FLJ46422
Gene Symbols COX6B2
Host Species Mouse
Immunogen Full length human recombinant protein of human COX6B2 (NP_653214) produced in HEK293T cell.
Molecular Weight of Antigen 10.3 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125965
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.