Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 154526
GREENER_CHOICESDP

Catalog No. 277910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-791-0 10 mg
27-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-791-0 Supplier Tocris Bioscience™ Supplier No. 2779/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, non-peptide CRF1 antagonist

Selective, non-peptide CRF1 receptor antagonist (Ki values are 2.7 and > 10000 nM for CRF1 and CRF2 receptors respectively). Blocks CRF-induced adenylate cyclase activation (Ki = 3.7 nM) and attenuates activation of the HPA axis by CRF. Displays anxiolytic-like activity in the rat elevated plus-maze test; brain penetrant.

Chemical Identifiers

CAS 257639-98-8
Molecular Formula C23H33ClN4

Specifications

Quantity 10 mg
Formula Weight 400.99
Purity >99%
Molecular Weight (g/mol) 400.995
Chemical Name or Material CP 154526
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.