Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 809101 hydrochloride
GREENER_CHOICESDP

Catalog No. 304110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-411-0 10 mg
30-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-411-0 Supplier Tocris Bioscience™ Supplier No. 3041/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

Potent and selective 5-HT2C receptor agonist (pEC50 values are 9.96, 7.19 and 6.81 for human 5-HT2C, 5-HT2B and 5-HT2A receptors respectively). Displays antipsychotic activity; suppresses condition avoidance responding (CAR) and inhibits PCP and amphetamine-stimulated hyperactivity in rats following subcutaneous administration.

Chemical Identifiers

CAS 1215721-40-6
Molecular Formula C15H18Cl2N4O

Specifications

Quantity 10 mg
Formula Weight 341.24
Purity >99%
Molecular Weight (g/mol) 341.236
Chemical Name or Material CP 809101 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.