Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CP 93129 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and highly selective 5-HT1B agonist (Ki values are 8.1, 1100, 1500, 2900 and 7200 nM for 5-HT1B, 5-HT1D, 5-HT1A, 5-HT1c and 5-HT2, respectively). Reduces food intake and decreases body weight in rats.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 288.18 |
Purity | >98% |
Molecular Weight (g/mol) | 306.187 |
Chemical Name or Material | CP 93129 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction