Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CP110 Mouse anti-Human, Clone: 1A3C12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CP110 |
---|---|
Clone | 1A3C12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6874
![]() ![]() |
Proteintech Group Inc
664481IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
CP110 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419 | |
CCP110 | |
IgG2a | |
Protein A | |
Antibody |
1A3C12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O43303 | |
9738 | |
CP110 Fusion Protein Ag3489 | |
Primary | |
-20°C | |
CCP110 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title