Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPEB1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358335 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358335 Supplier Novus Biologicals Supplier No. NBP176365MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPEB1 Polyclonal antibody specifically detects CPEB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen CPEB1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CPEBCEBP, CPE-binding protein 1, CPE-BP1, cytoplasmic polyadenylation element binding protein 1, cytoplasmic polyadenylation element-binding protein 1, FLJ13203, h-CEBP, hCPEB-1, MGC34136, MGC60106
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human CPEB1. The immunogen is located within amino acids 50 - 100 of CPEB1. Amino Acid Squence: LPDFQDSEETVTSR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 64506
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.