Learn More
MilliporeSigma™ Chemicon™ CpGenome™ Turbo Bisulfite Modification Kit
Catalog No. S7847 Shop All MilliporeSigma Products
Click to view available options
Quantity:
50 Reactions
Simplify and streamline bisulfite modification process
TRUSTED_SUSTAINABILITY
Specifications
Description CpGenome turbo bisulfite modification kit
For Use With (Application) Simplify and streamline bisulfite modification process
Includes Bisulfite conversion reagent 1, Bisulfite conversion reagent 2, Bisulfite reagent diluent, DNA binding buffer, DNA wash buffer, DNA elution buffer, modified DNA purification columns, 2mL collection tubes
Quantity 50 Reactions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.