Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPI17 alpha Antibody (4H10), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP104336AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104336AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104336AI Supplier Novus Biologicals Supplier No. NBP104336AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPI17 alpha Monoclonal specifically detects CPI17 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CPI17 alpha
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4H10
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Symbols PPP1R14A
Host Species Mouse
Immunogen Recombinant human PPP1R14A (1-147aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 94274
Test Specificity Anti-human PPP1R14A mAb, clone 4H10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPP1R14A protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.