Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPI17 alpha Antibody (4H10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310197 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310197 Supplier Novus Biologicals Supplier No. NBP104336MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPI17 alpha Monoclonal antibody specifically detects CPI17 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CPI17 alpha
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4H10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A
Host Species Mouse
Immunogen Recombinant human CPI17 alpha (1-147aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 94274
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.