Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPO Antibody (OTI1C1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272386A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272386A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272386A32 Supplier Novus Biologicals Supplier No. NBP272386AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPO Monoclonal specifically detects CPO in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CPO
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1C1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias carboxypeptidase O, EC 3.4.17, EC 3.4.17.-, MGC138281, MGC138283
Gene Symbols CPO
Host Species Mouse
Immunogen Full length recombinant protein of human CPO (NP_775100) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 130749
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.