Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPOX Antibody (36B10), PE, Novus Biologicals™
SDP

Catalog No. NB308971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308971 Supplier Novus Biologicals Supplier No. NBP259438PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPOX Monoclonal antibody specifically detects CPOX in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CPOX
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 36B10
Conjugate PE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Coprogen oxidase, coproporphyrinogen oxidase, coproporphyrinogenase, coproporphyrinogen-III oxidase, mitochondrial, COX, CPO, CPXcoproporphyrinogen oxidase (coproporphyria, harderoporphyria), EC 1.3.3.3, HCP
Host Species Mouse
Immunogen Recombinant human CPOX (111-454aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1371
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.