Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPR8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17075820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17075820 20 μL
NBP170758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17075820 Supplier Novus Biologicals Supplier No. NBP17075820UL

Rabbit Polyclonal Antibody

CPR8 Polyclonal specifically detects CPR8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CPR8
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Alias CCP8, cell cycle progression 1, cell cycle progression protein 1, cell cycle progression restoration protein 8, CPR8, FLJ60574, KIAA1254
Gene Symbols CCPG1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CCPG1(cell cycle progression 1) The peptide sequence was selected from the middle region of CCPG1. Peptide sequence TNLATENQYLRVSLEKEEKALSSLQEELNKLREQIRILEDKGTSTELVKE.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9236
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.