Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPS1 Antibody (CPS1/1022), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP247904A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247904A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247904A30 Supplier Novus Biologicals Supplier No. NBP247904AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPS1 Monoclonal specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CPS1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone CPS1/1022
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16
Gene Symbols CPS1
Host Species Mouse
Immunogen Recombinant human CPS1 protein (Uniprot: P31327)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1373
Test Specificity This MAb recognizes a protein of 165kDa, identified as carbamoyl phosphate synthetase 1 (CPS1). This mitochondrial enzyme catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells.Deficiency of CPS1 is an autosomal recessive disorder that causes hyperammonemia. CPS1 is a hepatocyte specific protein that localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. CPS1 may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of CPS1 correlates with smaller tumor size and longer patient survival. Occasionally, CPS1 is also found in gastric carcinomas as well as in a few other non-hepatic tumors.
Target Species Human, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.