Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPSF160 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501560745
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-156-0745 100 μL
1 options

Catalog No. 50-156-0745

Supplier: Bethyl Laboratories, Inc A301580A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Bovine

CPSF1 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that plays a key role in pre-mRNA 3′-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. This subunit is involved in the RNA recognition step of the polyadenylation reaction.

Specifications

Antigen CPSF160
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CPSF1
Gene Accession No. Q10570, Q9EPU4
Gene Alias Cleavage and polyadenylation specific factor 1, CPSF160, HSU37012, P/cl.18
Gene Symbols Cpsf1
Host Species Rabbit
Immunogen Between 1392 and 1442
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29894, 94230
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.