Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPSF6 Antibody, Novus Biologicals™
SDP

Catalog No. NB10061596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061596 Supplier Novus Biologicals Supplier No. NB10061596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

CPSF6 Polyclonal specifically detects CPSF6 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen CPSF6
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Simple Western 1:40, Immunohistochemistry 1:200 -1:1000, Immunocytochemistry/Immunofluorescence 1:50 to 1:500, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:200 -1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q16630
Gene Alias CFIM, CFIm68, CFIM68CPSF 68 kDa subunit, CFIMcleavage and polyadenylation specific factor 6, 68kD subunit, cleavage and polyadenylation specific factor 6, 68kDa, Cleavage and polyadenylation specificity factor 68 kDa subunit, cleavage and polyadenylation specificity factor subunit 6, HPBRII-4, HPBRII-7, pre-mRNA cleavage factor I, 68kD subunit, pre-mRNA cleavage factor Im (68kD), Pre-mRNA cleavage factor Im 68 kDa subunit, Protein HPBRII-4/7
Gene Symbols CPSF6
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cleavage and polyadenylation specific factor 6, 68kDa using the numbering given in entry NP_008938.1 (GeneID 11052).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11052
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Medaka fish, Nile tilapia, White-tufted-ear marmoset and Spotted green pufferfish.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.