Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPT1A Rabbit anti-Human, Mouse, Rat, Clone: 3M6E9, Novus Biologicals™
SDP

Catalog No. NB167239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167239 100 μg
NB167240 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167239 Supplier Novus Biologicals Supplier No. NBP316072100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CPT1A Monoclonal antibody specifically detects CPT1A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen CPT1A
Applications Western Blot
Classification Monoclonal
Clone 3M6E9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, Carnitine palmitoyltransferase 1A, carnitine palmitoyltransferase 1A (liver), carnitine palmitoyltransferase I, liver, CPT I, CPT1, CPT1-LEC 2.3.1.21, CPTI-L, EC 2.3.1, L-CPT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 520-720 of human CPT1A (NP_001867.2). WDIPGECQEVIETSLNTANLLANDVDFHSFPFVAFGKGIIKKCRTSPDAFVQLALQLAHYKDMGKFCLTYEASMTRLFREGRTETVRSCTTESCDFVRAMVDPAQTVEQRLKLFKLASEKHQHMYRLAMTGSGIDRHLFCLYVVSKYLAVESPFLKEVLSEPWRLSTSQTPQQQVELFDLENNPEYVSSGGGFGPVADDGY
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1374
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.