Learn More
CPT1C Mouse anti-Human, Porcine, Clone: 5D11B5, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 660721IG150UL

Description
This antibody can bind the close sequences genes.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
CPT1C | |
Monoclonal | |
0.98 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TCG5 | |
CPT1C | |
CPT1C Fusion Protein Ag17963 | |
150 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
5D11B5 | |
Unconjugated | |
CPT1C | |
CATL1, CPT IC, CPT1 B, CPT1C, CPT1P, CPTI B, CPTIC | |
Mouse | |
Protein A | |
RUO | |
100519258, 126129 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.