Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1C Mouse anti-Human, Porcine, Clone: 5D11B5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CPT1C |
---|---|
Clone | 5D11B5 |
Concentration | 0.98 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6509
![]() ![]() |
Proteintech Group Inc
660721IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody can bind the close sequences genes.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.Specifications
CPT1C | |
0.98 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CATL1, CPT IC, CPT1 B, CPT1C, CPT1P, CPTI B, CPTIC | |
CPT1C | |
IgG2b | |
Protein A | |
Antibody |
5D11B5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
Q8TCG5 | |
100519258, 126129 | |
CPT1C Fusion Protein Ag17963 | |
Primary | |
-20°C | |
CPT1C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title