Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRABP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120970 Supplier Invitrogen™ Supplier No. PA5120970
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, SW480, A-431, 22Rv1, HT-29, 293T, Mouse brain, Mouse uterus. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum, Nucleus. Immunogen sequence: VLGVNVMLRK IAVAAASKPA VEIKQEGDTF YIKTSTTVRT TEINFKVGEE FEEQTVDGRP CKSLVKWESE NKM.

Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRABP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRABP2
Gene Accession No. P22935, P29373, P51673
Gene Alias AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4
Gene Symbols CRABP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-94 of human CRABP2 (NP_001869.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12904, 1382, 29563
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.