Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRABP2 Rabbit anti-Human, Clone: 16, Novus Biologicals™
SDP

Catalog No. NB021144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021144 50 μg
NB021143 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021144 Supplier Novus Biologicals Supplier No. NBP28983750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CRABP2 Monoclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot.

Specifications

Antigen CRABP2
Applications Western Blot
Classification Monoclonal
Clone 16
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation 0.2 um filtered solution in PBS
Gene Alias cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6
Host Species Rabbit
Immunogen Recombinant Human CRABP2 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1382
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.