Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRACC/SLAMF7 Antibody (003), Alexa Fluor™ 594
SDP

Catalog No. NB074005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074005 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074005 Supplier Novus Biologicals Supplier No. NBP290477AF594

Rabbit Monoclonal Antibody

CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Mouse samples. It is validated for ELISA.

Specifications

Antigen CRACC/SLAMF7
Applications ELISA
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CRACC/SLAMF7 extracellular domain (NP_653122.2; Met 1-Gly 224).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 57823
Test Specificity Has cross-reactivity in ELISA with: Human SLAMF7/ CRACCM
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.