Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRACC/SLAMF7 Antibody (235614), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349495 Supplier Novus Biologicals Supplier No. FAB1906MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CRACC/SLAMF7
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 235614
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7, Lys27-Ser225, Accession # Q9NQ25
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 57823
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.