Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRALBP Antibody (OTI1G6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB271275APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271275APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271275APC Supplier Novus Biologicals Supplier No. NBP271275APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRALBP Monoclonal specifically detects CRALBP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CRALBP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G6
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1
Gene Symbols RLBP1
Host Species Mouse
Immunogen Full length recombinant protein of human CRALBP (NP_000317) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 6017.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.