Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CRANAD 2
SDP

Catalog No. 480310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-031-0 Supplier Tocris Bioscience™ Supplier No. 4803/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Near-infrared probe that detects Aβ40 aggregates

Near-infrared probe that binds to Aβ40 aggregates (Kd = 38 nM) and elicits an emission blue shift. Shown to bind to plaques in APP-PS1 transgenic mice, in vitro. Detects senile plaques in 19-month-old Tg2576 mice in vivo. Penetrates the blood-brain barrier.

Chemical Identifiers

CAS 1193447-34-5
Molecular Formula C23H25BF2N2O2

Specifications

Quantity 10 mg
Formula Weight 410.26
Purity >98%
Molecular Weight (g/mol) 410.272
Chemical Name or Material CRANAD 2
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.