Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRB2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB163799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163799 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB163799 Supplier Novus Biologicals Supplier No. NBP281710AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRB2 Polyclonal antibody specifically detects CRB2 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen CRB2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias crumbs homolog 2 (Drosophila)
Host Species Rabbit
Immunogen Rabbit polyclonal CRB2 antibody was raised against a 17 amino acid peptide near the center of human CRB2. The immunogen is located within amino acids 460 - 510 of CRB2. Amino Acid Squence: AGTLATRNDTKESLELA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286204
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.