Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRBN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598708 Supplier Invitrogen™ Supplier No. PA598708
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRBN Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRBN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CRBN
Gene Accession No. Q8C7D2, Q96SW2
Gene Alias 2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; cereblon; CRBN; DKFZp781K0715; MGC27358; MRT2; MRT2A; novel lethal; piL; protein cereblon; Protein PiL; protein x 0001
Gene Symbols CRBN
Host Species Rabbit
Immunogen Recombinant Human Protein cereblon protein (1-280AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51185, 58799
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.