Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cre Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB10056133X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056133X 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056133X Supplier Novus Biologicals Supplier No. NB10056133AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cre Polyclonal specifically detects Cre in Cre/lox System, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Cre
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cyclization recombinase
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of bacteriophage protein CRE (within amino acids 50-100). [UniProt# P06956]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2777477
Target Species Cre/lox System, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Target Molecule Cre/lox Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.