Learn More
Description
Specifications
Specifications
| Antigen | Cre |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | Cyclization recombinase |
| Host Species | Rabbit |
| Immunogen | Full-length recombinant bacteriophage protein CRE was used as immunogen, GenBank no. AAQ14086.1. |
| Molecular Weight of Antigen | 39 kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
