Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Kinase MB Antibody (1F2/1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062195 0.1 mg
NB017439 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062195 Supplier Novus Biologicals Supplier No. NB10062195
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Creatine Kinase MB Monoclonal specifically detects Creatine Kinase MB in Human samples. It is validated for ELISA, Radioimmune Assays (RIA).

Specifications

Antigen Creatine Kinase MB
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1F2/1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000
Gene Accession No. P06732, P12277
Gene Alias Creatine Kinase Isoenzyme
Gene Symbols CKB
Host Species Mouse
Immunogen Highly purified human CKMB.
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1152
Test Specificity Creatine phosphokinase, also known as creatine kinase (CK), is an enzyme expressed by various tissues and cell types. CK catalyses the conversion of creatine and consumes adenosine triphosphate (ATP) to create phosphocreatine and adenosine diphosphate (ADP). In cells, the “cytosolic” CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. NB100-62195 recognises CKMB but does not distinguish between isoform CKMB1 and CKMB2. This antibody shows less than 0.1% reactivity with CKBB or CKMM and minimal reactivity with other human serum proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.