Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Creatine Kinase MM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583885 Supplier Invitrogen™ Supplier No. PA583885
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPFGNTHNKF KLNYKPEEEY PDLS Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 92%.

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Creatine Kinase MM
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CKM
Gene Accession No. P06732
Gene Alias CKM; CKMM; CPK-M; Creatine kinase M chain; creatine kinase M-type; creatine kinase, M-type; creatine kinase, muscle; creatine phosphokinase M-type; M creatine phosphokinase; MCK; M-CK; muscle creatine kinase
Gene Symbols CKM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKM. Recombinant protein control fragment (Product #RP-100360).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1158
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.